Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029746.3 | NP_084022.2 | 8 | 944 | ||
GenBank mRNA | AK089920.1 | - | 8 | 928 | ||
AK157348.1 | - | 8 | 906 | |||
AK169298.1 | - | 8 | 914 | |||
AK171726.1 | - | 8 | 863 | |||
BC011435.1 | - | 8 | 884 | 4010102 | ||
CX205121.1 | - | 2 | 153 |