Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700028K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175241.1 | NP_780450.1 | 3, 4 | 287 | ||
NM_182745.2 | NP_877422.1 | 3, 4 | 287 | |||
GenBank mRNA | AK006463.1 | - | 3, 4 | 288 | ||
AK032886.1 | - | 4, 5 | 487 | |||
CN835533.1 | - | 3, 4 | 262 |