Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029619.2 | NP_083895.1 | 2, 3 | 244 | ||
GenBank mRNA | AK009912.1 | - | 2, 3 | 235 | ||
BC021619.1 | - | 2, 3 | 244 | 5150285 |