Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029749.2 | NP_084025.2 | 10 | 1140 | ||
GenBank mRNA | AK010801.1 | - | 9 | 950 | ||
AY336567.1 | - | 9 | 1013 | |||
BC137852.1 | - | 9 | 1033 | 8860871 | ||
BG079818.2 | - | 6 | 422 | |||
BM940384.1 | - | 2 | 55 | |||
CF532651.1 | - | 1 | 39 |