Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Catsper3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252487.1 | NP_001239416.1 | 7 | 1151 | ||
NM_001252488.1 | NP_001239417.1 | 7 | 998 | |||
NM_029772.4 | NP_084048.1 | 7 | 1112 | |||
GenBank mRNA | AK014942.1 | - | 7 | 1113 | ||
BC089518.1 | - | 1251 | 6774298 | |||
HM179540.1 | - | 7 | 1068 | |||
HM179541.1 | - | 7 | 915 |