Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178633.3 | NP_848748.2 | 874 | |||
GenBank mRNA | AK032116.1 | - | 422 | |||
AK167903.1 | - | 665 | ||||
AK220570.1 | - | 874 | ||||
BC024572.1 | - | 41 | ||||
BC031142.1 | - | 745 | 4972965 | |||
BC031144.1 | - | 768 | 4973325 |