Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161628.1 | NP_001155100.1 | 6, 7 | 950 | ||
NM_001161629.1 | NP_001155101.1 | 4, 5 | 438 | |||
NM_029946.4 | NP_084222.4 | 23, 24 | 3057 | |||
GenBank mRNA | AK016516.1 | - | 4, 5 | 543 | ||
AK019850.1 | - | 6, 7 | 950 | |||
AK220412.1 | - | 6, 7 | 869 | |||
BC056955.1 | - | 11, 12 | 1460 | 6812632 | ||
BC065111.1 | - | 23, 24 | 3057 | 30536704 |