Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175265.4 | NP_780474.1 | 3, 4 | 574 | ||
GenBank mRNA | AK034930.1 | - | 3, 4 | 575 | ||
AK141389.1 | - | 3, 4 | 525 | |||
AK167430.1 | - | 3, 4 | 506 | |||
BC052183.1 | - | 3, 4 | 475 | 6336038 |