Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023876.3 | NP_076365.2 | 4 | 470 | ||
GenBank mRNA | AJ276004.1 | - | 4 | 469 | ||
AK020742.1 | - | 4 | 478 | |||
AK034040.1 | - | 4 | 475 | |||
AK034529.1 | - | 4 | 476 | |||
AK037104.1 | - | 4 | 494 | |||
AK039447.1 | - | 4 | 482 | |||
AK039758.1 | - | 4 | 471 | |||
AK079069.1 | - | 4 | 485 | |||
AK168964.1 | - | 4 | 534 | |||
EF410086.1 | - | 334 |