Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081335.2 | NP_001074804.2 | 38 | 7245 | ||
GenBank mRNA | AK051474.1 | - | 15 | 2474 | ||
AK122351.1 | - | 35 | 6235 | |||
AK147675.1 | - | 36 | 7859 | |||
AK162968.1 | - | 17 | 2706 |