Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uvrag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178635.3 | NP_848750.3 | 15 | 1632 | ||
GenBank mRNA | AK030814.1 | - | 15 | 1563 | ||
AK043084.1 | - | 15 | 1610 | |||
AK078085.1 | - | 15 | 1624 | |||
AK154903.1 | - | 15 | 1632 | |||
AK165185.1 | - | 15 | 1613 | |||
BC034072.1 | - | 11 | 1348 | |||
BC034176.1 | - | 15 | 1601 | 4954871 |