Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172435.3 | NP_766023.1 | 3 | 1286 | ||
GenBank mRNA | AK020001.1 | - | 3 | 1165 | ||
AK038124.1 | - | 2 | 1213 | |||
AK041351.1 | - | 3 | 1302 | |||
AK088234.1 | - | 3 | 1287 | |||
BC053707.1 | - | 3 | 1117 | 30072257 |