Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163332.1 | NP_001156804.1 | 4 | 435 | ||
NM_001163333.1 | NP_001156805.1 | 4 | 427 | |||
NM_030249.4 | NP_084525.1 | 4 | 543 | |||
GenBank mRNA | AK029901.1 | - | 4 | 544 | ||
AK030438.1 | - | 4 | 408 | |||
AK051259.1 | - | 4 | 407 | |||
AK129359.1 | - | 4 | 433 | |||
AK145085.1 | - | 4 | 406 | |||
BC003236.1 | - | 4 | 397 | 3498078 | ||
CV562515.1 | - | 4 | 427 | |||
EF410090.1 | - | 404 |