Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mynn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289621.1 | NP_001276550.1 | 7 | 2201 | ||
NM_001289622.1 | NP_001276551.1 | 8 | 2155 | |||
NM_001289623.1 | NP_001276552.1 | 7 | 2071 | |||
NM_030557.3 | NP_085034.2 | 8 | 2285 | |||
GenBank mRNA | AF349561.1 | - | 7 | 1677 | ||
AK014315.1 | - | 7 | 1652 | |||
AK019238.2 | - | 8 | 1729 | |||
AK029657.1 | - | 8 | 1576 | |||
AK030898.1 | - | 7 | 1527 | |||
AK136107.1 | - | 8 | 1734 | |||
AK147959.1 | - | 7 | 1525 | |||
AK163261.1 | - | 8 | 1599 | |||
AK163264.1 | - | 8 | 1599 | |||
BC007477.1 | - | 8 | 1598 | |||
BC065084.1 | - | 8 | 2285 | 6848601 |