Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mynn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289621.1 | NP_001276550.1 | 2 | 845 | ||
NM_030557.3 | NP_085034.2 | 2 | 845 | |||
GenBank mRNA | AF349561.1 | - | 1 | 237 | ||
AK013445.1 | - | 2 | 300 | |||
AK014315.1 | - | 2 | 296 | |||
AK019238.2 | - | 2 | 289 | |||
AK136107.1 | - | 2 | 294 | |||
BC065084.1 | - | 2 | 845 | 6848601 |