Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141921.1 | NP_001135393.1 | 4 | 1063 | ||
NM_030562.2 | NP_085039.2 | 4 | 1024 | |||
GenBank mRNA | AB243740.1 | - | 1 | 845 | ||
AK090175.1 | - | 4 | 1064 | |||
AK144493.1 | - | 4 | 1065 | |||
BC004018.1 | - | 3 | 971 | 3494023 | ||
DQ070870.1 | - | 1 | 845 | |||
DQ314497.1 | - | 1 | 845 |