Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030564.1 | NP_085041.1 | 2 | 269 | ||
GenBank mRNA | AF434815.1 | - | 1 | 164 | ||
AK045696.1 | - | 2 | 265 | |||
AK046274.1 | - | 2 | 251 | |||
AK046381.1 | - | 2 | 265 | |||
AK077221.1 | - | 2 | 266 | |||
AK132330.1 | - | 285 | ||||
AK137606.1 | - | 245 | ||||
AK150976.1 | - | 2 | 263 | |||
AK159753.1 | - | 2 | 251 | |||
AK166565.1 | - | 278 | ||||
BC004042.1 | - | 2 | 269 | 3496314 |