Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159394.1 | NP_001152866.1 | 5 | 1069 | ||
NM_001159395.1 | NP_001152867.1 | 6 | 951 | |||
NM_030612.3 | NP_085115.1 | 7 | 1299 | |||
GenBank mRNA | AB020974.1 | - | 7 | 1511 | ||
AB026551.2 | - | 5 | 1069 | |||
AB047549.2 | - | 6 | 964 | |||
AK082908.1 | - | 5 | 1068 | |||
AK136509.1 | - | 2 | 538 | |||
AK141318.1 | - | 5 | 1070 | |||
AK149946.1 | - | 6 | 947 | |||
AK150519.1 | - | 5 | 1068 | |||
AK152016.1 | - | 5 | 1070 | |||
AK156826.1 | - | 5 | 1070 | |||
AK159093.1 | - | 5 | 1070 | |||
AK159171.1 | - | 5 | 1070 | |||
AK159459.1 | - | 5 | 1070 | |||
AK159645.1 | - | 5 | 1070 | |||
AK159917.1 | - | 5 | 1070 | |||
AK160065.1 | - | 5 | 1068 | |||
BC058188.1 | - | 5 | 1054 | 5341736 |