Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030701.3 | NP_109626.1 | 1 | 895 | ||
GenBank mRNA | AJ300198.1 | - | 1 | 875 | ||
AK150795.1 | - | 1 | 877 | |||
AK169956.1 | - | 1 | 880 | |||
BC120715.1 | - | 1 | 865 | 40129638 | ||
BC125296.1 | - | 1 | 865 | 40129811 |