Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030708.2 | NP_109633.2 | 10 | 6703 | ||
GenBank mRNA | AB024499.1 | - | 11 | 6234 |