Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023186.3 | NP_075675.2 | 11 | 1347 | ||
GenBank mRNA | AF154571.1 | - | 11 | 1341 | ||
AF290003.1 | - | 11 | 1346 | |||
AK007573.1 | - | 11 | 1355 | |||
AK008633.1 | - | 11 | 1360 | |||
AK008650.2 | - | 11 | 1358 | |||
AK008757.1 | - | 11 | 1360 | |||
AK160173.1 | - | 11 | 1360 | |||
BC011134.1 | - | 11 | 1342 | 4165150 | ||
BC034548.1 | - | 11 | 1347 | 4189080 | ||
DQ349202.1 | - | 11 | 1344 | |||
EF094027.1 | - | 11 | 1344 |