Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031185.3 | NP_112462.1 | 3 | 3428 | ||
GenBank mRNA | AB020886.1 | - | 3 | 3377 | ||
AB025278.1 | - | 3 | 3391 | |||
AB051563.1 | - | 3 | 3379 | |||
AB070852.1 | - | 2 | 3113 | |||
AB070853.1 | - | 2 | 3099 | |||
AF326228.1 | - | 3 | 3377 | |||
AF326230.1 | - | 2 | 3082 | |||
BC042461.1 | - | 1 | 2824 | |||
BC043939.1 | - | 1 | 2824 | |||
BC138364.1 | - | 3 | 3373 | 8861384 | ||
BC138365.1 | - | 3 | 3372 | 8861385 |