Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031251.4 | NP_112541.1 | 11 | 1183 | ||
GenBank mRNA | AJ250670.1 | - | 10 | 1081 | ||
AK054467.1 | - | 12 | 1361 | |||
AK078511.1 | - | 11 | 1195 | |||
AK079295.1 | - | 11 | 1184 | |||
AK085517.1 | - | 12 | 1365 | |||
AK137143.1 | - | 11 | 1226 | |||
AK170790.1 | - | 11 | 1195 | |||
AK170828.1 | - | 11 | 1206 | |||
BC005479.1 | - | 11 | 1170 | 3484952 | ||
BC006033.1 | - | 11 | 1170 |