Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032005.4 | NP_114394.1 | 1 | 367 | ||
GenBank mRNA | AF345520.1 | - | 1 | 193 | ||
AF348321.1 | - | 1 | 324 | |||
AK038666.1 | - | 1 | 369 | |||
BC120635.1 | - | 1 | 238 | 40129558 | ||
BC137731.1 | - | 1 | 238 | 8860750 |