Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpnmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053110.4 | NP_444340.3 | 3 | 523 | ||
GenBank mRNA | AF322054.1 | - | 3 | 361 | ||
AJ251685.1 | - | 3 | 408 | |||
AK044764.1 | - | 3 | 417 | |||
AK076347.1 | - | 3 | 392 | |||
AK079220.1 | - | 365 | ||||
AK149700.1 | - | 3 | 392 | |||
AK149740.1 | - | 3 | 430 | |||
AK150739.1 | - | 3 | 392 | |||
AK154617.1 | - | 3 | 427 | |||
AK159165.1 | - | 3 | 422 | |||
AK159270.1 | - | 3 | 430 | |||
AK159747.1 | - | 3 | 392 | |||
AK159908.1 | - | 3 | 398 | |||
AK170506.1 | - | 3 | 422 | |||
AK171347.1 | - | 3 | 398 | |||
AK171441.1 | - | 3 | 392 | |||
AK171465.1 | - | 3 | 392 | |||
AK171474.1 | - | 3 | 392 | |||
AK171517.1 | - | 3 | 510 | |||
AK171608.1 | - | 3 | 392 | |||
AK171618.1 | - | 3 | 392 | |||
AK190327.1 | - | 2 | 186 | |||
AK191573.1 | - | 1 | 59 | |||
AK192052.1 | - | 1 | 59 | |||
AK194062.1 | - | 1 | 59 | |||
AK194082.1 | - | 1 | 59 | |||
AK194155.1 | - | 1 | 59 | |||
AK197527.1 | - | 1 | 59 | |||
AK202122.1 | - | 1 | 59 | |||
AK202963.1 | - | 2 | 202 | |||
AK203550.1 | - | 1 | 60 | |||
AK204527.1 | - | 1 | 59 | |||
AK209686.1 | - | 1 | 59 | |||
BC026375.1 | - | 3 | 397 | 4164706 |