Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289530.1 | NP_001276459.1 | 6 | 1761 | ||
NM_052977.5 | NP_443209.2 | 6 | 1761 | |||
GenBank mRNA | AF232942.1 | - | 6 | 1378 | ||
AF270495.1 | - | 6 | 1478 | |||
AK033916.1 | - | 6 | 1700 | |||
AK045538.1 | - | 6 | 1700 | |||
AK046083.1 | - | 6 | 1705 | |||
AK134964.1 | - | 6 | 1763 | |||
AK139079.1 | - | 6 | 1719 | |||
AK139273.1 | - | 6 | 1719 | |||
BC052426.1 | - | 6 | 1686 | 6835942 | ||
BC059822.1 | - | 6 | 1714 | 6844585 |