Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033325.2 | NP_201582.2 | 13 | 2359 | ||
GenBank mRNA | AF117951.1 | - | 556 | |||
AK034957.1 | - | 13 | 2397 | |||
AK045019.1 | - | 13 | 2359 | |||
AK145323.1 | - | 13 | 2397 | |||
AK159386.1 | - | 13 | 2393 | |||
BC086801.1 | - | 2641 | 30605240 |