Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030131.3 | NP_084407.1 | 2 | 166 | ||
GenBank mRNA | AK021301.1 | - | 2 | 120 | ||
AK144157.1 | - | 2 | 127 | |||
AK165179.1 | - | 2 | 112 | |||
AK166153.1 | - | 2 | 110 | |||
BC040509.1 | - | 2 | 106 | |||
BC065395.1 | - | 2 | 110 | 876056 |