Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027874.2 | NP_082150.1 | 6 | 1117 | ||
NM_139059.2 | NP_620690.1 | 6 | 1117 | |||
GenBank mRNA | AK004606.1 | - | 6 | 877 | ||
AK051052.1 | - | 6 | 1116 | |||
AK088642.1 | - | 6 | 852 | |||
AK140317.1 | - | 6 | 842 | |||
AK143534.1 | - | 6 | 818 | |||
AK146600.1 | - | 6 | 1107 | |||
AK150387.1 | - | 6 | 846 | |||
AK150815.1 | - | 6 | 844 | |||
AK152721.1 | - | 6 | 1059 | |||
AK157812.1 | - | 6 | 852 | |||
AK190835.1 | - | 1 | 20 | |||
BC004604.1 | - | 6 | 1103 | 3585286 |