Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146061.4 | NP_666173.4 | 1, 2 | 317 | ||
GenBank mRNA | AF482997.1 | - | 1, 2 | 312 | ||
AK046057.1 | - | 2, 3 | 134 | |||
AK081231.1 | - | 1, 2 | 313 | |||
BC024991.1 | - | 2, 3 | 201 | 5065129 | ||
BY059925.1 | - | 1, 2 | 289 |