Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tirap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177845.1 | NP_001171316.1 | 6 | 595 | ||
NM_001177846.1 | NP_001171317.1 | 6 | 577 | |||
NM_001177847.1 | NP_001171318.1 | 6 | 523 | |||
NM_054096.2 | NP_473437.2 | 6 | 587 | |||
GenBank mRNA | AF378130.1 | - | 6 | 496 | ||
AF410784.1 | - | 5 | 419 | |||
AK036431.1 | - | 6 | 516 | |||
AK080135.1 | - | 6 | 596 | |||
AK151117.1 | - | 6 | 525 | |||
AK168494.1 | - | 6 | 515 | |||
BC005584.1 | - | 3 | 253 | 3157913 | ||
BY743490.1 | - | 6 | 523 |