Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007556.2 | NP_031582.1 | 3 | 1116 | ||
GenBank mRNA | AK041210.1 | - | 1474 | |||
AK135770.1 | - | 3 | 749 | |||
BC138593.1 | - | 3 | 1036 | 8861613 | ||
BC138595.1 | - | 3 | 1036 | 8861615 | ||
J04566.1 | - | 3 | 698 | |||
X80992.1 | - | 3 | 1115 |