Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp36l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007564.5 | NP_031590.1 | 2 | 736 | ||
GenBank mRNA | AK036950.1 | - | 2 | 737 | ||
AK051035.1 | - | 2 | 737 | |||
AK077855.1 | - | 2 | 733 | |||
BC016621.1 | - | 2 | 721 | 3157001 | ||
M58566.1 | - | 2 | 665 |