Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cited1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276466.1 | NP_001263395.1 | 2, 3 | 279 | ||
NM_001276473.1 | NP_001263402.1 | 2, 3 | 148 | |||
NM_001276474.1 | NP_001263403.1 | 2, 3 | 395 | |||
NM_007709.4 | NP_031735.1 | 2, 3 | 482 | |||
GenBank mRNA | AK148042.1 | - | 2, 3 | 261 | ||
BC052030.1 | - | 2, 3 | 408 | 5709775 | ||
BQ173936.1 | - | 634 | ||||
CX237826.1 | - | 2, 3 | 394 | |||
U65091.1 | - | 2, 3 | 261 |