Search Thermo Fisher Scientific
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012266.1 | - | 2, 3 | 462 | ||
AK027977.1 | - | 2, 3 | 434 | |||
BC004620.1 | - | 2, 3 | 440 | 3585323 | ||
BC004703.1 | - | 2, 3 | 1072 | 3499265 | ||
DV661455.1 | - | 1, 2 | 36 | |||
X97755.1 | - | 2, 3 | 438 |