Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012266.1 | - | 5 | 651 | ||
AK027977.1 | - | 5 | 623 | |||
BC004620.1 | - | 5 | 629 | 3585323 | ||
BC004703.1 | - | 5 | 1261 | 3499265 | ||
DV661455.1 | - | 4 | 225 | |||
X97755.1 | - | 5 | 627 |