Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007924.2 | NP_031950.2 | 8 | 1309 | ||
GenBank mRNA | AK159027.1 | - | 8 | 1296 | ||
BC014816.1 | - | 8 | 1279 | 4482339 | ||
BC024894.1 | - | 8 | 1278 | 4159557 | ||
U80227.1 | - | 8 | 1282 |