Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136062.2 | NP_001129534.1 | 8 | 862 | ||
NM_001276285.1 | NP_001263214.1 | 8 | 874 | |||
NM_007933.3 | NP_031959.1 | 8 | 974 | |||
GenBank mRNA | AK002485.1 | - | 8 | 834 | ||
BC013460.1 | - | 7 | 846 | 4217678 | ||
CT010193.1 | - | 7 | 768 | |||
M20745.1 | - | 5 | 594 | |||
X57747.1 | - | 8 | 825 | |||
X62667.1 | - | 7 | 818 |