Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111066.1 | NP_001104536.1 | 7 | 1058 | ||
NM_001199631.1 | NP_001186560.1 | 7 | 1154 | |||
NM_010223.2 | NP_034353.2 | 7 | 1055 | |||
GenBank mRNA | AB154247.1 | - | 6 | 794 | ||
AF030635.1 | - | 6 | 825 | |||
AK088739.1 | - | 7 | 993 | |||
AK146122.1 | - | 7 | 1053 | |||
AK152198.1 | - | 7 | 1054 | |||
AK153000.1 | - | 7 | 1051 | |||
AK165281.1 | - | 7 | 1051 | |||
AK167663.1 | - | 7 | 1041 | |||
AY225340.1 | - | 6 | 938 | |||
AY278608.1 | - | 7 | 1014 | |||
BC003739.1 | - | 6 | 974 | 3601511 | ||
BC027808.1 | - | 7 | 1008 | 5324380 |