Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008057.3 | NP_032083.3 | 1 | 1824 | ||
GenBank mRNA | BC049781.1 | - | 1 | 1547 | ||
BC063077.1 | - | 2 | 1155 | 6844727 | ||
U43320.1 | - | 1 | 1372 |
Set Membership: |
![]() |