Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gpx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008161.3 | NP_032187.2 | 3 | 415 | ||
GenBank mRNA | AK002219.1 | - | 3 | 344 | ||
AK002262.1 | - | 3 | 344 | |||
AK004942.1 | - | 3 | 344 | |||
AK146760.1 | - | 3 | 344 | |||
BC003339.1 | - | 3 | 327 | 3257333 | ||
BC037027.1 | - | 3 | 362 | 5370640 | ||
BC037134.1 | - | 3 | 341 | |||
BC049235.1 | - | 3 | 331 | 4221025 | ||
BC061950.1 | - | 3 | 319 | 6528998 | ||
CB601893.1 | - | 3 | 347 | |||
U13705.1 | - | 3 | 319 |