Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gstm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160411.1 | NP_001153883.1 | 4 | 312 | ||
NM_026764.3 | NP_081040.1 | 5 | 517 | |||
GenBank mRNA | AF464943.1 | - | 5 | 347 | ||
AF501320.1 | - | 5 | 333 | |||
AK003418.1 | - | 4 | 313 | |||
BC030444.1 | - | 5 | 464 | 5373062 | ||
CK335113.1 | - | 5 | 502 |