Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177301.5 | NP_796275.3 | 3 | 518 | ||
GenBank mRNA | AK044748.1 | - | 3 | 294 | ||
AK085906.1 | - | 3 | 469 | |||
AK144684.1 | - | 3 | 472 | |||
BC027206.1 | - | 3 | 495 | 4218471 | ||
BC099683.1 | - | 3 | 513 | 6530863 |