- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Kcnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112739.2 | NP_001106210.1 | 2 | 2242 | ||
NM_008421.3 | NP_032447.1 | 2 | 2242 | |||
GenBank mRNA | AK147476.1 | - | 2 | 2215 | ||
AK147682.1 | - | 2 | 2215 | |||
AK162964.1 | - | 2 | 899 | |||
BC125470.1 | - | 2 | 1079 | 40129985 | ||
BC132439.1 | - | 2 | 1079 | 40130716 | ||
Y07521.1 | - | 2 | 1168 |