Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008448.3 | NP_032474.2 | 19 | 2533 | ||
GenBank mRNA | BC090841.1 | - | 19 | 2509 | 30543821 | |
L27153.1 | - | 19 | 2127 | |||
U86090.1 | - | 19 | 2360 |