Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008451.2 | NP_032477.2 | 9 | 1464 | ||
GenBank mRNA | AF055666.1 | - | 9 | 1297 | ||
AK141439.1 | - | 9 | 1394 | |||
BC014845.1 | - | 9 | 1308 | 2599082 | ||
BM963360.1 | - | 5 | 635 | |||
CX565742.1 | - | 1 | 64 |