Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Psmb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010724.2 | NP_034854.2 | 5 | 892 | ||
GenBank mRNA | AK031801.1 | - | 5 | 717 | ||
AK050627.1 | - | 5 | 720 | |||
AK146182.1 | - | 5 | 739 | |||
AK150674.1 | - | 5 | 720 | |||
AK159971.1 | - | 5 | 709 | |||
AK169753.1 | - | 3 | 1604 | |||
BC013785.1 | - | 5 | 716 | 2654130 | ||
BC051450.1 | - | 5 | 723 | 30076739 | ||
KJ650228.1 | - | 5 | 688 | |||
U22031.1 | - | 5 | 688 | |||
U22032.1 | - | 5 | 688 | |||
U22033.1 | - | 5 | 688 | |||
U22034.1 | - | 5 | 688 | |||
U22035.1 | - | 5 | 688 | |||
X64449.1 | - | 5 | 707 |