Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010776.1 | NP_034906.1 | 6 | 706 | ||
GenBank mRNA | AK149484.1 | - | 6 | 736 | ||
BC010760.1 | - | 6 | 721 | 4212216 | ||
D11440.1 | - | 6 | 706 | |||
S42294.1 | - | 6 | 732 |