Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008562.3 | NP_032588.1 | 2 | 934 | ||
GenBank mRNA | AK010424.1 | - | 1 | 109 | ||
AK159504.1 | - | 2 | 846 | |||
AK160594.1 | - | 1 | 106 | |||
AK215970.1 | - | 1 | 159 | |||
BC003839.1 | - | 2 | 863 | 3491015 | ||
BC005427.1 | - | 2 | 856 | 3493739 | ||
BC021638.1 | - | 2 | 873 | 4190996 | ||
GU182318.1 | - | 3 | 660 | |||
U35623.1 | - | 2 | 846 |