Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cited2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010828.3 | NP_034958.2 | 2 | 521 | ||
GenBank mRNA | AK131664.1 | - | 2 | 524 | ||
AK133791.1 | - | 2 | 522 | |||
BC057126.1 | - | 2 | 503 | 6415181 | ||
U86445.1 | - | 2 | 501 | |||
Y15163.1 | - | 2 | 500 |